Product Details

SNP ID
rs142752193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37007537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTGGTCCACTGGAACAGTCAGG[A/T]GTTTTTGTAGCATCTTCTAAGTCTG
Phenotype
MIM: 116957
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RBL1 PubMed Links

Gene Details

Gene
RBL1
Gene Name
RB transcriptional corepressor like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323281.1 2808 Silent Mutation ACA,ACT T488T NP_001310210.1
NM_001323282.1 2808 Silent Mutation ACA,ACT T488T NP_001310211.1
NM_002895.4 2808 Silent Mutation ACA,ACT T915T NP_002886.2
NM_183404.3 2808 Silent Mutation ACA,ACT T915T NP_899662.1
XM_006723842.2 2808 Silent Mutation ACA,ACT T743T XP_006723905.1
XM_011528955.2 2808 Silent Mutation ACA,ACT T915T XP_011527257.1
XM_011528958.2 2808 Intron XP_011527260.1
XM_017027992.1 2808 Silent Mutation ACA,ACT T811T XP_016883481.1

View Full Product Details