Product Details

SNP ID
rs143135336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2794369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCGTGTCACTGAATGGTAGCCCT[C/T]GGCACTGGCAGTCACCATGTAGTCC
Phenotype
MIM: 609555
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPXM1 PubMed Links

Gene Details

Gene
CPXM1
Gene Name
carboxypeptidase X (M14 family), member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184699.1 1878 Missense Mutation AAG,GAG K602E NP_001171628.1
NM_019609.4 1878 Missense Mutation AAG,GAG K676E NP_062555.1

View Full Product Details