Product Details

SNP ID
rs143245012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:25454781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGCACTGCTGCACGTCAGTTCC[C/T]ACCTGCTGAGTTGGACTTCGAACAT
Phenotype
MIM: 610608 MIM: 609580
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GINS1 PubMed Links

Gene Details

Gene
GINS1
Gene Name
GINS complex subunit 1
There are no transcripts associated with this gene.

Gene
NINL
Gene Name
ninein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318226.1 Intron NP_001305155.1
NM_025176.5 Intron NP_079452.3
XM_011529186.2 Intron XP_011527488.1
XM_011529187.2 Intron XP_011527489.1
XM_011529188.1 Intron XP_011527490.1
XM_011529189.1 Intron XP_011527491.1
XM_011529190.1 Intron XP_011527492.1
XM_011529191.1 Intron XP_011527493.1
XM_011529192.1 Intron XP_011527494.1
XM_011529193.1 Intron XP_011527495.1
XM_011529194.2 Intron XP_011527496.1
XM_011529195.2 Intron XP_011527497.1
XM_011529197.2 Intron XP_011527499.1
XM_011529198.1 Intron XP_011527500.1
XM_017027732.1 Intron XP_016883221.1
XM_017027733.1 Intron XP_016883222.1
XM_017027734.1 Intron XP_016883223.1
XM_017027735.1 Intron XP_016883224.1

View Full Product Details