Product Details

SNP ID
rs143468590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31844686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGTTGGCAGGGGCAGTGGGGCC[C/T]GGAGTGAGCCTGGAGGGGTGGGTGG
Phenotype
MIM: 616776 MIM: 602939 MIM: 606566
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP15 PubMed Links

Gene Details

Gene
DUSP15
Gene Name
dual specificity phosphatase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012644.2 932 Intron NP_001012662.1
NM_001320478.1 932 Intron NP_001307407.1
NM_001320479.1 932 Intron NP_001307408.1
NM_080611.4 932 Intron NP_542178.2
NM_177991.2 932 Intron NP_817130.1
XM_017027654.1 932 Intron XP_016883143.1
XM_017027655.1 932 Intron XP_016883144.1
XM_017027656.1 932 Intron XP_016883145.1
XM_017027657.1 932 Intron XP_016883146.1
XM_017027658.1 932 Intron XP_016883147.1
XM_017027659.1 932 Intron XP_016883148.1
XM_017027660.1 932 Intron XP_016883149.1
XM_017027661.1 932 Intron XP_016883150.1
XM_017027662.1 932 Intron XP_016883151.1
Gene
FOXS1
Gene Name
forkhead box S1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004118.3 932 Missense Mutation CAG,CGG Q286R NP_004109.1
Gene
MYLK2
Gene Name
myosin light chain kinase 2
There are no transcripts associated with this gene.

View Full Product Details