Product Details
- SNP ID
-
rs143478494
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:46177716 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGCTAGGGTGAGGTTTGGGCTACA[C/T]GTGAATGGGGTGGGCCACAGTGAGC
- Phenotype
-
MIM: 609920
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDH22
PubMed Links
Gene Details
- Gene
- CDH22
- Gene Name
- cadherin 22
View Full Product Details