Product Details
- SNP ID
-
rs143498763
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:63645553 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCTGTGCCCTGTCAGGGGAAGAG[A/G]AAGGCGCCTGCTGTCACAGTGAAAA
- Phenotype
-
MIM: 608362
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
STMN3
PubMed Links
Gene Details
- Gene
- STMN3
- Gene Name
- stathmin 3
View Full Product Details