Product Details

SNP ID
rs144014784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31844861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGTAGGGGCCTTATTAAAACTCT[C/T]AGCCTCTGAGAAGCCGGCAGGAAAG
Phenotype
MIM: 616776 MIM: 602939
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP15 PubMed Links

Gene Details

Gene
DUSP15
Gene Name
dual specificity phosphatase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012644.2 757 Intron NP_001012662.1
NM_001320478.1 757 Intron NP_001307407.1
NM_001320479.1 757 Intron NP_001307408.1
NM_080611.4 757 Intron NP_542178.2
NM_177991.2 757 Intron NP_817130.1
XM_017027654.1 757 Intron XP_016883143.1
XM_017027655.1 757 Intron XP_016883144.1
XM_017027656.1 757 Intron XP_016883145.1
XM_017027657.1 757 Intron XP_016883146.1
XM_017027658.1 757 Intron XP_016883147.1
XM_017027659.1 757 Intron XP_016883148.1
XM_017027660.1 757 Intron XP_016883149.1
XM_017027661.1 757 Intron XP_016883150.1
XM_017027662.1 757 Intron XP_016883151.1
Gene
FOXS1
Gene Name
forkhead box S1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004118.3 757 Missense Mutation AAG,GAG K228E NP_004109.1

View Full Product Details