Product Details

SNP ID
rs144347230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63644281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGAGCAGATGAGCGACAGCAC[C/T]GACAGCTCCTTCATCTTCTCCTTGT
Phenotype
MIM: 608362
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STMN3 PubMed Links

Gene Details

Gene
STMN3
Gene Name
stathmin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276310.1 313 Silent Mutation TCA,TCG S5S NP_001263239.1
NM_015894.3 313 Silent Mutation TCA,TCG S16S NP_056978.2

View Full Product Details