Product Details

SNP ID
rs144586846
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3228365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGCAGCTGAAGCACCTGCAGGC[C/T]CGTGAAGTAGTGGATCTTCCTCTGG
Phenotype
MIM: 147520 MIM: 610206
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ITPA PubMed Links
Additional Information
For this assay, SNP(s) [rs58757394] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ITPA
Gene Name
inosine triphosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267623.1 2575 Intron NP_001254552.1
NM_001324236.1 2575 Intron NP_001311165.1
NM_001324237.1 2575 Intron NP_001311166.1
NM_001324238.1 2575 Intron NP_001311167.1
NM_001324240.1 2575 Intron NP_001311169.1
NM_033453.3 2575 Intron NP_258412.1
NM_181493.3 2575 Intron NP_852470.1
XM_006723564.3 2575 Intron XP_006723627.1
XM_006723565.3 2575 Intron XP_006723628.1
XM_011529234.2 2575 Intron XP_011527536.1
Gene
SLC4A11
Gene Name
solute carrier family 4 member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174089.1 2575 Missense Mutation AGC,GGC S818G NP_001167560.1
NM_001174090.1 2575 Missense Mutation AGC,GGC S861G NP_001167561.1
NM_032034.3 2575 Missense Mutation AGC,GGC S834G NP_114423.1
XM_005260856.4 2575 Missense Mutation AGC,GGC S941G XP_005260913.1
XM_005260857.1 2575 Missense Mutation AGC,GGC S799G XP_005260914.1
XM_011529383.2 2575 Missense Mutation AGC,GGC S807G XP_011527685.1
XM_011529384.1 2575 Missense Mutation AGC,GGC S799G XP_011527686.1
XM_011529385.1 2575 Missense Mutation AGC,GGC S799G XP_011527687.1
XM_017028093.1 2575 Missense Mutation AGC,GGC S939G XP_016883582.1
XM_017028094.1 2575 Missense Mutation AGC,GGC S799G XP_016883583.1
XM_017028095.1 2575 Missense Mutation AGC,GGC S780G XP_016883584.1
XM_017028096.1 2575 Missense Mutation AGC,GGC S799G XP_016883585.1
XM_017028097.1 2575 Intron XP_016883586.1

View Full Product Details