Product Details

SNP ID
rs145079188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:20052554 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGGCTGCGCGTCCTCCTTGCGG[C/T]AGCGCGTGGAGTGCGGCGTCCTGGA
Phenotype
MIM: 610952
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFAP61 PubMed Links

Gene Details

Gene
CFAP61
Gene Name
cilia and flagella associated protein 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167816.1 304 Intron NP_001161288.1
NM_015585.3 304 UTR 5 NP_056400.3
XM_005260688.3 304 UTR 5 XP_005260745.1
XM_005260689.2 304 UTR 5 XP_005260746.1
XM_005260690.3 304 UTR 5 XP_005260747.1
XM_005260691.3 304 UTR 5 XP_005260748.1
XM_005260692.3 304 UTR 5 XP_005260749.1
XM_005260693.3 304 UTR 5 XP_005260750.1
XM_005260695.4 304 Intron XP_005260752.1
XM_005260696.4 304 Intron XP_005260753.1
XM_005260697.1 304 UTR 5 XP_005260754.1
XM_011529211.1 304 UTR 5 XP_011527513.1
XM_011529212.2 304 UTR 5 XP_011527514.1
XM_011529213.1 304 UTR 5 XP_011527515.1
XM_017027786.1 304 UTR 5 XP_016883275.1
XM_017027787.1 304 UTR 5 XP_016883276.1
XM_017027788.1 304 UTR 5 XP_016883277.1
XM_017027789.1 304 UTR 5 XP_016883278.1
XM_017027790.1 304 UTR 5 XP_016883279.1
XM_017027791.1 304 Intron XP_016883280.1
XM_017027792.1 304 Intron XP_016883281.1
XM_017027793.1 304 Intron XP_016883282.1
XM_017027794.1 304 Intron XP_016883283.1
XM_017027795.1 304 UTR 5 XP_016883284.1
Gene
CRNKL1
Gene Name
crooked neck pre-mRNA splicing factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278625.1 304 Missense Mutation TAC,TGC Y79C NP_001265554.1
NM_001278626.1 304 UTR 5 NP_001265555.1
NM_001278627.1 304 UTR 5 NP_001265556.1
NM_001278628.1 304 Intron NP_001265557.1
NM_016652.5 304 Missense Mutation TAC,TGC Y91C NP_057736.4

View Full Product Details