Product Details

SNP ID
rs145112384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31377421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAATTCCACTGTTACCCATGCAT[C/T]GAAGGATGTGGCGTTTGCCTGCCAA
Phenotype
MIM: 607650 MIM: 615997
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEFB118 PubMed Links

Gene Details

Gene
DEFB118
Gene Name
defensin beta 118
There are no transcripts associated with this gene.

Gene
DEFB119
Gene Name
defensin beta 119
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271209.1 246 Intron NP_001258138.1
NM_153289.3 246 Missense Mutation CAA,CGA Q27R NP_695021.2
NM_153323.4 246 Intron NP_697018.1

View Full Product Details