Product Details

SNP ID
rs145235171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34560930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCAAGTAGAGGCCATGTGTGAGG[C/T]AGTACTCCCGCCGCAGGAAGGCATA
Phenotype
MIM: 601242 MIM: 608528
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP1LC3A PubMed Links

Gene Details

Gene
MAP1LC3A
Gene Name
microtubule associated protein 1 light chain 3 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032514.3 1444 Intron NP_115903.1
NM_181509.2 1444 Intron NP_852610.1
XM_011529083.2 1444 Intron XP_011527385.1
XM_011529084.2 1444 Intron XP_011527386.1
XM_011529085.2 1444 Intron XP_011527387.1
Gene
PIGU
Gene Name
phosphatidylinositol glycan anchor biosynthesis class U
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080476.4 1444 Missense Mutation TAC,TGC Y415C NP_536724.1
XM_011528542.2 1444 Missense Mutation TAC,TGC Y199C XP_011526844.1
XM_017027664.1 1444 Missense Mutation TAC,TGC Y367C XP_016883153.1
XM_017027665.1 1444 Intron XP_016883154.1

View Full Product Details