Product Details

SNP ID
rs145631155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63305966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGGAGCCCACCCCCTCCCACAC[A/G]GGGAGCCCAGACCCTGAGCAGGCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL20A1 PubMed Links

Gene Details

Gene
COL20A1
Gene Name
collagen type XX alpha 1 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020882.2 540 Silent Mutation ACA,ACG T141T NP_065933.2
XM_011528937.1 540 Silent Mutation ACA,ACG T141T XP_011527239.1
XM_011528938.1 540 Silent Mutation ACA,ACG T141T XP_011527240.1
XM_011528939.1 540 Silent Mutation ACA,ACG T141T XP_011527241.1
XM_011528940.1 540 Silent Mutation ACA,ACG T141T XP_011527242.1

View Full Product Details