Product Details

SNP ID
rs146505168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:25616137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATCACACAGTCCCCAGGTTGTA[C/T]TCCGAGAAGATTGCAGCAGTAATAA
Phenotype
MIM: 610763
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NANP PubMed Links

Gene Details

Gene
NANP
Gene Name
N-acetylneuraminic acid phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152667.2 599 Missense Mutation ATA,GTA I179V NP_689880.1
Gene
ZNF337-AS1
Gene Name
ZNF337 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details