Product Details

SNP ID
rs147861977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1273427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCTGGCCACGGTGAAGGAATTC[A/C]AAGAAGTATTTACAACCATTCATTC
Phenotype
MIM: 604942
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SNPH PubMed Links

Gene Details

Gene
SNPH
Gene Name
syntaphilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318234.1 Intron NP_001305163.1
NM_014723.3 Intron NP_055538.2
XM_005260889.3 Intron XP_005260946.1
XM_011529402.2 Intron XP_011527704.1
XM_011529403.2 Intron XP_011527705.1
XM_011529404.2 Intron XP_011527706.1

View Full Product Details