Product Details

SNP ID
rs148522459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63643841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTGGGTGGGGAGGAGAGCATA[C/G]GGCTCTCTGGGGACAGGTCAGAAGG
Phenotype
MIM: 608362
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
STMN3 PubMed Links

Gene Details

Gene
STMN3
Gene Name
stathmin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276310.1 471 Missense Mutation CCT,CGT P58R NP_001263239.1
NM_015894.3 471 Missense Mutation CCT,CGT P69R NP_056978.2

View Full Product Details