Product Details

SNP ID
rs149390633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:61254848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCCCATAATGAGGATCTTACAA[C/T]TAGAGAGACCTGCAAGGCTGGGTTC
Phenotype
MIM: 603006
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDH4 PubMed Links

Gene Details

Gene
CDH4
Gene Name
cadherin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252338.2 168 Intron NP_001239267.1
NM_001252339.2 168 Intron NP_001239268.1
NM_001794.4 168 Missense Mutation ACT,ATT T27I NP_001785.2

View Full Product Details