Product Details
- SNP ID
-
rs150159953
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:44337273 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGCCCCGGCCCAGCCCGAGAGCA[C/T]GGCTATGCGGCTCCTGAGTGGCCTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
R3HDML
PubMed Links
Gene Details
- Gene
- R3HDML
- Gene Name
- R3H domain containing like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178491.3 |
116 |
Missense Mutation |
ACG,ATG |
T39M |
NP_848586.1 |
View Full Product Details