Product Details

SNP ID
rs151034191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:5938466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGATATGAAAAAACAAGTAGTAA[C/T]GGCATTTAAAGTTTAATTACTAACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRMT6 PubMed Links

Gene Details

Gene
TRMT6
Gene Name
tRNA methyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281467.1 1812 UTR 3 NP_001268396.1
NM_015939.4 1812 UTR 3 NP_057023.2
XM_017027875.1 1812 UTR 3 XP_016883364.1

View Full Product Details