Product Details

SNP ID
rs139822824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:20743762 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAGATGACCAGTTTCTCAGTGT[C/T]TAAAGTTCATCTTCTTGCTGAGCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00320 PubMed Links

Gene Details

Gene
LINC00320
Gene Name
long intergenic non-protein coding RNA 320
There are no transcripts associated with this gene.

View Full Product Details