Product Details
- SNP ID
-
rs140909597
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:41740855 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGCCAGCTAGGTCTTGCTTCGCC[A/G]CAGGAGCGTGGCGGCGGGGCCATGG
- Phenotype
-
MIM: 605706
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR6814
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7276592] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR6814
- Gene Name
- microRNA 6814
There are no transcripts associated with this gene.
- Gene
- RIPK4
- Gene Name
- receptor interacting serine/threonine kinase 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020639.2 |
2386 |
Missense Mutation |
CGG,TGG |
R780W |
NP_065690.2 |
View Full Product Details