Product Details

SNP ID
rs142352513
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:17598771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTGGAGATCTCATCCTTGTTC[C/T]CATCTTTATTTTCAAAGCTGGCAAC
Phenotype
MIM: 605674 MIM: 602621
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTG3 PubMed Links

Gene Details

Gene
BTG3
Gene Name
BTG anti-proliferation factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130914.1 720 Missense Mutation GAG,GGG E166G NP_001124386.1
NM_006806.4 720 Missense Mutation GAG,GGG E122G NP_006797.3
XM_011529441.1 720 Missense Mutation GAG,GGG E176G XP_011527743.1
XM_017028265.1 720 Missense Mutation GAG,GGG E122G XP_016883754.1
XM_017028266.1 720 Missense Mutation GAG,GGG E122G XP_016883755.1
Gene
CXADR
Gene Name
coxsackie virus and adenovirus receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207063.1 720 Intron NP_001193992.1
NM_001207064.1 720 Intron NP_001193993.1
NM_001207065.1 720 Intron NP_001193994.1
NM_001207066.1 720 Intron NP_001193995.1
NM_001338.4 720 Intron NP_001329.1
XM_011529476.2 720 Intron XP_011527778.1
XM_011529477.2 720 Intron XP_011527779.1
XM_011529478.2 720 Intron XP_011527780.1
XM_011529479.1 720 Intron XP_011527781.1

View Full Product Details