Product Details

SNP ID
rs142713888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30501994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCCACCGCCATAGCCCAGTCTGC[A/G]GAAGCTGCCACATCCACAGCCATAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP19-4 PubMed Links

Gene Details

Gene
KRTAP19-4
Gene Name
keratin associated protein 19-4
There are no transcripts associated with this gene.

Gene
KRTAP19-5
Gene Name
keratin associated protein 19-5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181611.1 97 Missense Mutation CGC,TGC R33C NP_853642.1

View Full Product Details