Product Details

SNP ID
rs143198588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33436866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACTCAGCCCATCTTAGAGGCCTT[A/G]GACAAGGACAGCTCACCAAAGGATG
Phenotype
MIM: 147569
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFNGR2 PubMed Links

Gene Details

Gene
IFNGR2
Gene Name
interferon gamma receptor 2 (interferon gamma transducer 1)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005534.3 1566 Silent Mutation TTA,TTG L306L NP_005525.2
XM_005260969.2 1566 Silent Mutation TTA,TTG L325L XP_005261026.1
XM_011529553.1 1566 Silent Mutation TTA,TTG L331L XP_011527855.1
XM_011529554.2 1566 Silent Mutation TTA,TTG L308L XP_011527856.1
Gene
TMEM50B
Gene Name
transmembrane protein 50B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006134.6 1566 Intron NP_006125.2
XM_011529746.1 1566 Intron XP_011528048.1

View Full Product Details