Product Details
- SNP ID
-
rs144905070
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:30371789 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTTGGGCGGCAGAATCCAGATCCA[C/T]AGCCGAGGGAAGGGAAGCCACAGCT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KRTAP13-2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs877346] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRTAP13-2
- Gene Name
- keratin associated protein 13-2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181621.3 |
451 |
Missense Mutation |
TAT,TGT |
Y142C |
NP_853652.1 |
- Gene
- MIR4327
- Gene Name
- microRNA 4327
There are no transcripts associated with this gene.
View Full Product Details