Product Details

SNP ID
rs145879341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30501941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGCTGCGGTATCCATAGCCTCC[A/G]AAGCCAGAGCCGTATCCGTAGCCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP19-3 PubMed Links

Gene Details

Gene
KRTAP19-3
Gene Name
keratin associated protein 19-3
There are no transcripts associated with this gene.

Gene
KRTAP19-4
Gene Name
keratin associated protein 19-4
There are no transcripts associated with this gene.

Gene
KRTAP19-5
Gene Name
keratin associated protein 19-5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181611.1 150 Silent Mutation TTC,TTT F50F NP_853642.1

View Full Product Details