Product Details

SNP ID
rs146717052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44456910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGGGGCCTTCCAGCACCTGCACC[A/G]GCTCAGGGAGCTGGATCTGTCTCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC3 PubMed Links

Gene Details

Gene
LRRC3
Gene Name
leucine rich repeat containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030891.4 583 Missense Mutation CAG,CGG Q89R NP_112153.1
Gene
LRRC3-AS1
Gene Name
LRRC3 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details