Product Details

SNP ID
rs147242429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44558082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGACTTCTGGCCTGAGGAGAGGC[C/T]GCAGCACGCGGAAGAGAGGCGGGAG
Phenotype
MIM: 612920
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP10-2 PubMed Links

Gene Details

Gene
KRTAP10-2
Gene Name
keratin associated protein 10-2
There are no transcripts associated with this gene.

Gene
KRTAP10-3
Gene Name
keratin associated protein 10-3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198696.2 Intron NP_941969.2
Gene
TSPEAR
Gene Name
thrombospondin type laminin G domain and EAR repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272037.1 Intron NP_001258966.1
NM_144991.2 Intron NP_659428.2

View Full Product Details