Product Details

SNP ID
rs147971699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44353850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCTGGAGGCTACAGTGCCCCTT[C/T]GGCAACAATGACAAGGTAGGCTTTC
Phenotype
MIM: 603749
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRPM2 PubMed Links

Gene Details

Gene
TRPM2
Gene Name
transient receptor potential cation channel subfamily M member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320350.1 655 Silent Mutation TTC,TTT F50F NP_001307279.1
NM_001320351.1 655 Silent Mutation TTC,TTT F50F NP_001307280.1
NM_001320352.1 655 Intron NP_001307281.1
NM_003307.3 655 Silent Mutation TTC,TTT F50F NP_003298.1
XM_005261171.3 655 Missense Mutation TTC,TTT F50F XP_005261228.1
XM_011529736.2 655 Missense Mutation TTC,TTT F50F XP_011528038.1
XM_017028456.1 655 Missense Mutation TTC,TTT F50F XP_016883945.1
XM_017028457.1 655 Missense Mutation TTC,TTT F50F XP_016883946.1

View Full Product Details