Product Details
- SNP ID
-
rs150331955
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:42315364 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACCAGCCCCAGCATGCAGAGCGCT[C/G]TGGCAGCCATGACCACCGTGGGCTC
- Phenotype
-
MIM: 600633
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TFF3
PubMed Links
Gene Details
- Gene
- TFF3
- Gene Name
- trefoil factor 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003226.3 |
233 |
Missense Mutation |
ACA,AGA |
T18R |
NP_003217.3 |
View Full Product Details