Product Details

SNP ID
rs150635388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30371744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGTCTGTAACAAGGAGACTGG[A/C]AGCTCCTAGAAGCCAAGTAGGTTGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP13-2 PubMed Links

Gene Details

Gene
KRTAP13-2
Gene Name
keratin associated protein 13-2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181621.3 496 Missense Mutation TGC,TTC C157F NP_853652.1
Gene
MIR4327
Gene Name
microRNA 4327
There are no transcripts associated with this gene.

View Full Product Details