Product Details

SNP ID
rs150850006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29085843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACAACTTGAGTGGTTAAATTACT[A/G]TCATGCAAAGCGACTAGATGGTTCA
Phenotype
MIM: 611110
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP3K7CL PubMed Links

Gene Details

Gene
MAP3K7CL
Gene Name
MAP3K7 C-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286617.1 274 UTR 5 NP_001273546.1
NM_001286618.1 274 UTR 5 NP_001273547.1
NM_001286619.1 274 Intron NP_001273548.1
NM_001286620.1 274 Intron NP_001273549.1
NM_001286622.1 274 UTR 5 NP_001273551.1
NM_001286623.1 274 UTR 5 NP_001273552.1
NM_001286624.1 274 Intron NP_001273553.1
NM_001286634.1 274 UTR 5 NP_001273563.1
NM_020152.3 274 UTR 5 NP_064537.1

View Full Product Details