Product Details

SNP ID
rs137966537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38294120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGGCAGCTCACCTGTGAGGCTG[A/G]GATCCGGGAGACCTCTTGCCGCCCA
Phenotype
MIM: 600863
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSNK1E PubMed Links

Gene Details

Gene
CSNK1E
Gene Name
casein kinase 1 epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001894.4 1318 Missense Mutation CCA,TCA P403S NP_001885.1
NM_152221.2 1318 Missense Mutation CCA,TCA P403S NP_689407.1
Gene
LOC400927-CSNK1E
Gene Name
LOC400927-CSNK1E readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289912.1 1318 Missense Mutation CCA,TCA P403S NP_001276841.1

View Full Product Details