Product Details

SNP ID
rs138618678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30264023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCCCCCACTTGCTGAAGACCC[A/G]CCCCACTGAGTGCATGAAGCGATGG
Phenotype
MIM: 165095
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OSM PubMed Links

Gene Details

Gene
OSM
Gene Name
oncostatin M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319108.1 663 Missense Mutation CGG,TGG R186W NP_001306037.1
NM_020530.5 663 Missense Mutation CGG,TGG R207W NP_065391.1

View Full Product Details