Product Details

SNP ID
rs139548825
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50265367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCTTGGCCTCAACGCTCTCATC[A/G]TATGGCTCGGCCTCTGGCTCATCCT
Phenotype
MIM: 602898 MIM: 602399 MIM: 604293
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAPK11 PubMed Links

Gene Details

Gene
MAPK11
Gene Name
mitogen-activated protein kinase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002751.6 794 Silent Mutation TAC,TAT Y323Y NP_002742.3
XM_011530706.1 794 Silent Mutation TAC,TAT Y215Y XP_011529008.1
Gene
MAPK12
Gene Name
mitogen-activated protein kinase 12
There are no transcripts associated with this gene.

Gene
PLXNB2
Gene Name
plexin B2
There are no transcripts associated with this gene.

View Full Product Details