Product Details

SNP ID
rs139943188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30379609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCCAGCTGGGCAGCTAATCATC[A/G]GAGTAGCGGTTCCCTGCAGGGGAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC157 PubMed Links

Gene Details

Gene
CCDC157
Gene Name
coiled-coil domain containing 157
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017437.3 1125 Intron NP_001017437.2
NM_001318334.1 1125 Intron NP_001305263.1
NM_001318335.1 1125 Intron NP_001305264.1
XM_006724278.3 1125 Intron XP_006724341.1
XM_006724279.3 1125 Intron XP_006724342.1
XM_006724280.3 1125 Intron XP_006724343.1
XM_011530263.2 1125 Intron XP_011528565.1
XM_011530266.2 1125 Intron XP_011528568.1
XM_011530267.2 1125 Intron XP_011528569.1
XM_011530268.2 1125 Intron XP_011528570.1
XM_011530269.2 1125 Intron XP_011528571.1
XM_011530270.2 1125 Intron XP_011528572.1
Gene
KIAA1656
Gene Name
KIAA1656 protein
There are no transcripts associated with this gene.

Gene
RNF215
Gene Name
ring finger protein 215
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017981.1 1125 Silent Mutation TCC,TCT S375S NP_001017981.1

View Full Product Details