Product Details

SNP ID
rs141833363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42086202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACTTCATGGATCGTGGCCAACA[C/T]AGAACTTGGATAGGAAATCCTTTGG
Phenotype
MIM: 614240 MIM: 602138 MIM: 615588
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM109B PubMed Links

Gene Details

Gene
FAM109B
Gene Name
family with sequence similarity 109 member B
There are no transcripts associated with this gene.

Gene
NDUFA6
Gene Name
NADH:ubiquinone oxidoreductase subunit A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002490.3 508 Missense Mutation TAT,TGT Y149C NP_002481.2
Gene
NDUFA6-AS1
Gene Name
NDUFA6 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
SMDT1
Gene Name
single-pass membrane protein with aspartate rich tail 1
There are no transcripts associated with this gene.

View Full Product Details