Product Details

SNP ID
rs141887267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20242171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTCCTCATCGGTGGCCCTGCCG[A/G]TCCAGATGGGATGGTAAGGGCCGGT
Phenotype
MIM: 605566
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR1286 PubMed Links

Gene Details

Gene
MIR1286
Gene Name
microRNA 1286
There are no transcripts associated with this gene.

Gene
RTN4R
Gene Name
reticulon 4 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023004.5 1163 Missense Mutation NP_075380.1

View Full Product Details