Product Details

SNP ID
rs141934594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50580386 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGATGTCATTGTGGCAGAAGAC[A/G]ACTGGCGATGGGGTAGACTCTAGTA
Phenotype
MIM: 612395 MIM: 601987
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHKB PubMed Links

Gene Details

Gene
CHKB
Gene Name
choline kinase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005198.4 926 Silent Mutation GTC,GTT V236V NP_005189.2
Gene
CHKB-AS1
Gene Name
CHKB antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
CHKB-CPT1B
Gene Name
CHKB-CPT1B readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
CPT1B
Gene Name
carnitine palmitoyltransferase 1B
There are no transcripts associated with this gene.

View Full Product Details