Product Details

SNP ID
rs142837500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19441162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAACCACCAGTCAGGCTGACAGAT[A/G]GTTTTCAACACCACAGGACTCCCTC
Phenotype
MIM: 600237 MIM: 605089 MIM: 601754
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C22orf39 PubMed Links

Gene Details

Gene
C22orf39
Gene Name
chromosome 22 open reading frame 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166242.1 1269 UTR 3 NP_001159714.1
NM_173793.4 1269 Intron NP_776154.3
Gene
HIRA
Gene Name
histone cell cycle regulator
There are no transcripts associated with this gene.

Gene
MRPL40
Gene Name
mitochondrial ribosomal protein L40
There are no transcripts associated with this gene.

Gene
UFD1L
Gene Name
ubiquitin fusion degradation 1 like (yeast)
There are no transcripts associated with this gene.

View Full Product Details