Product Details

SNP ID
rs143139801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50625391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCTTGGACAGGTCATAGAGCAG[C/T]GGGGGCTCATGAGCAGTCAGAGAGC
Phenotype
MIM: 607574
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARSA PubMed Links

Gene Details

Gene
ARSA
Gene Name
arylsulfatase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000487.5 1553 Silent Mutation CCA,CCG P428P NP_000478.3
NM_001085425.2 1553 Silent Mutation CCA,CCG P428P NP_001078894.2
NM_001085426.2 1553 Silent Mutation CCA,CCG P428P NP_001078895.2
NM_001085427.2 1553 Silent Mutation CCA,CCG P428P NP_001078896.2
NM_001085428.2 1553 Silent Mutation CCA,CCG P342P NP_001078897.1
XM_011530690.2 1553 Silent Mutation CCA,CCG P342P XP_011528992.1
XM_011530691.2 1553 UTR 3 XP_011528993.1
XM_017028800.1 1553 Silent Mutation CCA,CCG P466P XP_016884289.1

View Full Product Details