Product Details

SNP ID
rs143924347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:49993597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCAGGTTCCTCCCTTGTCTGGG[A/C]GGGACAGTAGGACAGCTGAGATCTG
Phenotype
MIM: 613414
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IL17REL PubMed Links

Gene Details

Gene
IL17REL
Gene Name
interleukin 17 receptor E like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001694.2 Intron NP_001001694.2
XM_011530689.2 Intron XP_011528991.1
XM_017028798.1 Intron XP_016884287.1

View Full Product Details