Product Details

SNP ID
rs144432807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:21634079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCGGCCAGCCTGCAAGCCGGGC[C/T]GTGTGCCACACCCACCATCCACATG
Phenotype
MIM: 607551
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC116 PubMed Links

Gene Details

Gene
CCDC116
Gene Name
coiled-coil domain containing 116
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152612.2 442 Missense Mutation CGT,TGT R44C NP_689825.2
XM_006724159.2 442 Missense Mutation CGT,TGT R110C XP_006724222.1
XM_011529984.2 442 Missense Mutation CGT,TGT R9C XP_011528286.1
XM_011529985.1 442 Missense Mutation CGT,TGT R110C XP_011528287.1
Gene
SDF2L1
Gene Name
stromal cell derived factor 2 like 1
There are no transcripts associated with this gene.

Gene
YDJC
Gene Name
YdjC homolog (bacterial)
There are no transcripts associated with this gene.

View Full Product Details