Product Details

SNP ID
rs144887903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:45193085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTCAAGGGCACGTGCCTTTCCT[C/T]ATCTCGTGGAGTGTCCGTCCTATCA
Phenotype
MIM: 604646
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA0930 PubMed Links

Gene Details

Gene
KIAA0930
Gene Name
KIAA0930
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009880.1 5416 UTR 3 NP_001009880.1
NM_015264.1 5416 UTR 3 NP_056079.1
XM_011530017.2 5416 UTR 3 XP_011528319.1
XM_017028672.1 5416 UTR 3 XP_016884161.1
Gene
MIR1249
Gene Name
microRNA 1249
There are no transcripts associated with this gene.

Gene
NUP50
Gene Name
nucleoporin 50
There are no transcripts associated with this gene.

View Full Product Details