Product Details

SNP ID
rs145032337
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:17633104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTAGAAAATACAAAAAAGTACA[A/G]AGAATAATTTATTCCCATACCATGC
Phenotype
MIM: 108746
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6V1E1 PubMed Links

Gene Details

Gene
ATP6V1E1
Gene Name
ATPase H+ transporting V1 subunit E1
There are no transcripts associated with this gene.

Gene
BCL2L13
Gene Name
BCL2 like 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270726.1 Intron NP_001257655.1
NM_001270727.1 Intron NP_001257656.1
NM_001270728.1 Intron NP_001257657.1
NM_001270729.1 Intron NP_001257658.1
NM_001270730.1 Intron NP_001257659.1
NM_001270731.1 Intron NP_001257660.1
NM_001270732.1 Intron NP_001257661.1
NM_001270733.1 Intron NP_001257662.1
NM_001270734.1 Intron NP_001257663.1
NM_001270735.1 Intron NP_001257664.1
NM_015367.3 Intron NP_056182.2
XM_011546119.1 Intron XP_011544421.1
XM_011546120.1 Intron XP_011544422.1
XM_011546121.2 Intron XP_011544423.1
XM_011546123.1 Intron XP_011544425.1
XM_017028725.1 Intron XP_016884214.1
XM_017028726.1 Intron XP_016884215.1
XM_017028727.1 Intron XP_016884216.1
XM_017028728.1 Intron XP_016884217.1
XM_017028729.1 Intron XP_016884218.1
XM_017028730.1 Intron XP_016884219.1
XM_017028731.1 Intron XP_016884220.1
XM_017028732.1 Intron XP_016884221.1
XM_017028733.1 Intron XP_016884222.1
XM_017028734.1 Intron XP_016884223.1

View Full Product Details