Product Details

SNP ID
rs145999812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38683815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTCAGGAGGAATGCCAGGGGCT[C/T]TACCCTCACTTCCTGGAAAGATCTA
Phenotype
MIM: 607757 MIM: 615323 MIM: 607046
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CBY1 PubMed Links

Gene Details

Gene
CBY1
Gene Name
chibby family member 1, beta catenin antagonist
There are no transcripts associated with this gene.

Gene
JOSD1
Gene Name
Josephin domain containing 1
There are no transcripts associated with this gene.

Gene
TOMM22
Gene Name
translocase of outer mitochondrial membrane 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020243.4 433 Silent Mutation CTA,TTA L135L NP_064628.1

View Full Product Details