Product Details
- SNP ID
-
rs146115534
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:42512916 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACTGCGGCTCTCACAGCTCAGTA[C/T]GGTCGGAATTTGCGCTGGGCCCGCA
- Phenotype
-
MIM: 607979
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RRP7A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9745] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RRP7A
- Gene Name
- ribosomal RNA processing 7 homolog A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015703.4 |
874 |
Silent Mutation |
|
|
NP_056518.2 |
- Gene
- SERHL
- Gene Name
- serine hydrolase-like (pseudogene)
There are no transcripts associated with this gene.
View Full Product Details