Product Details
- SNP ID
-
rs146350825
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:23692143 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGTCTGTGGGACGCCAGGGCGCA[C/T]GAGGGTCTGTACAGCCCTGCTGTAT
- Phenotype
-
MIM: 612214
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GUSBP11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17003394] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GUSBP11
- Gene Name
- glucuronidase, beta pseudogene 11
There are no transcripts associated with this gene.
- Gene
- RGL4
- Gene Name
- ral guanine nucleotide dissociation stimulator like 4
View Full Product Details