Product Details

SNP ID
rs146476554
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43113859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTACCGTCTGTGCTCAGAACAGTT[G/T]GAGCCGTATGGAGGAAGTCTCCGCA
Phenotype
MIM: 603392
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
BIK PubMed Links

Gene Details

Gene
BIK
Gene Name
BCL2 interacting killer
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197.4 Intron NP_001188.1

View Full Product Details