Product Details

SNP ID
rs147831812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:25201434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGAGCACCCGAACAGGCTGCAG[C/G]TGGCAAGAGCCATGGAGACCTTGGG
Phenotype
MIM: 123630
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CRYBB3 PubMed Links

Gene Details

Gene
CRYBB3
Gene Name
crystallin beta B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004076.4 656 Missense Mutation GCT,GGT A13G NP_004067.1
XM_011529902.2 656 Missense Mutation GCT,GGT A69G XP_011528204.1
XM_017028599.1 656 Missense Mutation GCT,GGT A69G XP_016884088.1
Gene
KIAA1671
Gene Name
KIAA1671
There are no transcripts associated with this gene.

View Full Product Details